siRNA Details
Gene Symbol: MAPKBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128608.1 | NP_001122080.1 | 12, 13 | 1613 | ||
NM_001265611.1 | NP_001252540.1 | 11, 12 | 1595 | |||
NM_014994.2 | NP_055809.2 | 11, 12 | 1595 | |||
NR_049761.1 | 10, 11 | 1405 | ||||
XM_011521384.2 | XP_011519686.1 | 12, 13 | 1577 | |||
GenBank mRNA | AB011168.2 | - | 12, 13 | 1523 | ||
AB177850.1 | - | 3, 4 | 443 | |||
AK091616.1 | - | 9, 10 | 2265 | |||
AL833267.1 | - | 11, 12 | 1515 | |||
BC036660.1 | - | 10, 11 | 1410 | 5266613 | ||
BC113983.1 | - | 11, 12 | 1487 | 40083186 | ||
BC114493.1 | - | 12, 13 | 1505 | 40083182 | ||
BX648786.1 | - | 11, 12 | 1546 |
