siRNA Details
Gene Symbol: CCDC152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134848.1 | NP_001128320.1 | 8 | 656 | ||
GenBank mRNA | AK127368.1 | - | 8 | 646 | ||
AK303469.1 | - | 6 | 488 | |||
BC039102.1 | - | 8 | 628 |
