siRNA Details
Gene Symbol: RILPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319243.1 | NP_001306172.1 | 6 | 1228 | ||
NM_001319244.1 | NP_001306173.1 | 6 | 777 | |||
NM_001319302.1 | NP_001306231.1 | 7 | 1727 | |||
NM_178314.4 | NP_847884.2 | 6 | 1228 | |||
XM_006719372.2 | XP_006719435.1 | 6 | 1222 | |||
XM_006719373.3 | XP_006719436.1 | 5 | 575 | |||
XM_011538282.2 | XP_011536584.1 | 6 | 1263 | |||
XM_011538284.2 | XP_011536586.1 | 5 | 794 | |||
XM_011538285.2 | XP_011536587.1 | 5 | 555 | |||
GenBank mRNA | AK096697.1 | - | 7 | 1070 | ||
BC080626.1 | - | 6 | 777 | 6339887 | ||
BC089444.1 | - | 6 | 1030 | 30559974 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
