siRNA Details
Gene Symbol: NEIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256552.1 | NP_001243481.1 | 3 | 883 | ||
NM_024608.3 | NP_078884.2 | 3 | 959 | |||
NR_046311.1 | 4 | 1082 | ||||
XM_005254659.4 | XP_005254716.1 | 3 | 819 | |||
XM_006720677.3 | XP_006720740.1 | 3 | 804 | |||
XM_006720679.3 | XP_006720742.1 | 3 | 742 | |||
XM_006720680.1 | XP_006720743.1 | 4 | 974 | |||
XM_006720681.1 | XP_006720744.1 | 4 | 821 | |||
XM_011522001.2 | XP_011520303.2 | 3 | 724 | |||
XM_011522002.1 | XP_011520304.1 | 4 | 947 | |||
XM_011522003.2 | XP_011520305.1 | 5 | 1003 | |||
XM_011522004.2 | XP_011520306.1 | 3 | 691 | |||
XM_011522005.1 | XP_011520307.1 | 4 | 1033 | |||
XM_011522006.1 | XP_011520308.1 | 3 | 957 | |||
XM_011522007.1 | XP_011520309.1 | 4 | 1053 | |||
XM_011522008.1 | XP_011520310.1 | 4 | 1026 | |||
XM_017022567.1 | XP_016878056.1 | 3 | 717 | |||
XM_017022568.1 | XP_016878057.1 | 3 | 841 | |||
GenBank mRNA | AB079068.1 | - | 2 | 453 | ||
AK026055.1 | - | 4 | 880 | |||
AK097008.1 | - | 4 | 1082 | |||
AK128372.1 | - | 4 | 554 | |||
BC010876.1 | - | 3 | 864 | 3879198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
