siRNA Details
Gene Symbol: TRAPPC3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139444.2 | NP_001132916.1 | 5 | 590 | ||
GenBank mRNA | AK002042.1 | - | 3 | 404 | ||
BC127757.1 | - | 2 | 321 | 40127548 | ||
BC127758.1 | - | 2 | 320 | 40127555 |
