siRNA Details
Gene Symbol: NPIPB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135865.1 | NP_001129337.1 | 7 | 2130 | ||
GenBank mRNA | AK293635.1 | - | 1 | 1088 | ||
AK296338.1 | - | 7 | 1971 | |||
AK303264.1 | - | 5 | 951 | |||
AK303572.1 | - | 1 | 1512 | |||
AK316332.1 | - | 1 | 378 |
Gene Symbol: LOC613037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321892.1 | NP_001308821.1 | 9 | 2059 | ||
XM_017023514.1 | XP_016879003.1 | 9 | 2348 | |||
XM_017023515.1 | XP_016879004.1 | 10 | 2367 | |||
XM_017023516.1 | XP_016879005.1 | 9 | 2394 | |||
XM_017023517.1 | XP_016879006.1 | 9 | 2125 | |||
XM_017023518.1 | XP_016879007.1 | 9 | 2213 | |||
XM_017023519.1 | XP_016879008.1 | 9 | 2270 | |||
XM_017023520.1 | XP_016879009.1 | 4 | 3334 | |||
XM_017023521.1 | XP_016879010.1 | 5 | 1589 | |||
XM_017023522.1 | XP_016879011.1 | 2 | 1998 |
Gene Symbol: NPIPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130464.2 | NP_569731.2 | 8 | 2053 | ||
GenBank mRNA | AB209632.1 | - | 9 | 3888 | ||
AK128772.1 | - | 4 | 2924 | |||
AK294452.1 | - | 4 | 1616 | |||
AK295170.1 | - | 9 | 1582 | |||
AK300277.1 | - | 1496 | ||||
AK303166.1 | - | 1 | 1260 | |||
AK303500.1 | - | 6 | 1673 | |||
AK304162.1 | - | 6 | 1823 | |||
AK316459.1 | - | 6 | 1608 |
Gene Symbol: NPIPB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310148.1 | NP_001297077.1 | 8 | 2217 | ||
XM_017023244.1 | XP_016878733.1 | 7 | 4228 | |||
XM_017023245.1 | XP_016878734.1 | 8 | 4065 | |||
XM_017023246.1 | XP_016878735.1 | 8 | 3986 | |||
XM_017023247.1 | XP_016878736.1 | 8 | 2093 | |||
GenBank mRNA | AK303607.1 | - | 1239 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
