siRNA Details
Gene Symbol: C19orf60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100418.1 | NP_001093888.1 | 5 | 854 | ||
NM_001100419.1 | NP_001093889.1 | 5 | 788 | |||
XM_005259977.1 | XP_005260034.1 | 3 | 775 | |||
GenBank mRNA | AK000857.1 | - | 5 | 742 | ||
AK054670.1 | - | 4 | 1946 | |||
AK091925.1 | - | 3 | 2250 | |||
AK094710.1 | - | 1 | 2441 | |||
BC012078.1 | - | 5 | 743 | 4661993 | ||
BI836487.1 | - | 5 | 665 | |||
KP143545.1 | - | 5 | 648 |
