siRNA Details
Gene Symbol: C14orf80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134875.1 | NP_001128347.1 | 7 | 998 | ||
NM_001134876.1 | NP_001128348.1 | 6 | 899 | |||
NM_001134877.1 | NP_001128349.1 | 7 | 826 | |||
NM_001198983.1 | NP_001185912.1 | 7 | 908 | |||
GenBank mRNA | AK024506.1 | - | 1 | 3740 | ||
BC016028.1 | - | 6 | 666 | 3907551 | ||
BX161477.1 | - | 7 | 908 | |||
BX248038.1 | - | 7 | 826 | |||
BX248074.1 | - | 8 | 1126 | |||
BX248283.1 | - | 6 | 857 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
