siRNA Details
Gene Symbol: TBC1D2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015079.5 | NP_055894.6 | 12, 13 | 2693 | ||
GenBank mRNA | AB028978.1 | - | 12, 13 | 2556 | ||
BC152465.1 | - | 12, 13 | 2556 | 8862601 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
