siRNA Details
Gene Symbol: TBC1D2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015079.5 | NP_055894.6 | 9 | 2200 | ||
NM_144572.1 | NP_653173.1 | 9 | 2200 | |||
XM_011521387.2 | XP_011519689.1 | 8 | 2145 | |||
XM_017022022.1 | XP_016877511.1 | 11 | 2221 | |||
GenBank mRNA | AB028978.1 | - | 9 | 2063 | ||
AK000173.1 | - | 2 | 1167 | |||
BC033712.1 | - | 4 | 983 | |||
BC140766.1 | - | 8 | 1845 | 9021634 | ||
BC144482.1 | - | 8 | 1845 | 9053009 | ||
BC152465.1 | - | 9 | 2063 | 8862601 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
