siRNA Details
Gene Symbol: TBC1D2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015079.5 | NP_055894.6 | 6 | 1265 | ||
NM_144572.1 | NP_653173.1 | 6 | 1265 | |||
XM_011521387.2 | XP_011519689.1 | 6 | 1321 | |||
XM_017022022.1 | XP_016877511.1 | 8 | 1286 | |||
GenBank mRNA | AB028978.1 | - | 6 | 1128 | ||
BC033712.1 | - | 1 | 48 | |||
BC140766.1 | - | 5 | 910 | 9021634 | ||
BC144482.1 | - | 5 | 910 | 9053009 | ||
BC152465.1 | - | 6 | 1128 | 8862601 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
