siRNA Details
Gene Symbol: TMEM87A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110503.2 | NP_001103973.1 | 2 | 379 | ||
NM_015497.4 | NP_056312.2 | 2 | 379 | |||
XM_005254287.1 | XP_005254344.1 | 2 | 230 | |||
XM_006720468.2 | XP_006720531.1 | 2 | 235 | |||
XM_011521444.2 | XP_011519746.1 | 2 | 230 | |||
XM_017022067.1 | XP_016877556.1 | 2 | 235 | |||
GenBank mRNA | AF132733.1 | - | 2 | 235 | ||
AK074870.1 | - | 2 | 228 | |||
AK075403.1 | - | 2 | 265 | |||
AK289591.1 | - | 2 | 228 | |||
AK290150.1 | - | 2 | 214 | |||
AK297816.1 | - | 2 | 224 | |||
AK309915.1 | - | 2 | 228 | |||
BC005335.1 | - | 2 | 231 | |||
DC341748.1 | - | 2 | 352 | |||
HY036655.1 | - | 2 | 379 |
