siRNA Details
Gene Symbol: SERPINA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000354.5 | NP_000345.2 | 3 | 816 | ||
XM_005262180.3 | XP_005262237.1 | 3 | 814 | |||
XM_006724683.2 | XP_006724746.1 | 3 | 814 | |||
GenBank mRNA | AK310778.1 | - | 3 | 814 | ||
BC020747.2 | - | 2 | 785 | 4716693 | ||
M14091.1 | - | 2 | 1099 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
