siRNA Details
Gene Symbol: ZNF680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178558.4 | NP_848653.2 | 4 | 713 | ||
XM_006715959.2 | XP_006716022.1 | 5 | 989 | |||
XM_017012126.1 | XP_016867615.1 | 7 | 1295 | |||
XM_017012127.1 | XP_016867616.1 | 6 | 1107 | |||
XM_017012128.1 | XP_016867617.1 | 6 | 1177 | |||
XM_017012129.1 | XP_016867618.1 | 5 | 839 | |||
GenBank mRNA | AK122937.1 | - | 4 | 674 | ||
BC030700.1 | - | 4 | 697 | 4837196 |
