siRNA Details
Gene Symbol: ZNF680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130022.1 | NP_001123494.1 | 2 | 300 | ||
NM_178558.4 | NP_848653.2 | 2 | 300 | |||
XM_006715959.2 | XP_006716022.1 | 3 | 576 | |||
XM_017012126.1 | XP_016867615.1 | 3 | 576 | |||
XM_017012127.1 | XP_016867616.1 | 3 | 576 | |||
XM_017012128.1 | XP_016867617.1 | 3 | 576 | |||
XM_017012129.1 | XP_016867618.1 | 2 | 308 | |||
GenBank mRNA | AK122937.1 | - | 2 | 261 | ||
AK131240.1 | - | 2 | 214 | |||
BC030700.1 | - | 2 | 284 | 4837196 | ||
DA669757.1 | - | 2 | 300 |
