siRNA Details
Gene Symbol: CTAGE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198495.2 | NP_940897.2 | 1 | 2341 | ||
GenBank mRNA | AF338232.1 | - | 1 | 1666 | ||
DB515636.2 | - | 1 | 434 |
Gene Symbol: CTAGE15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC146818.1 | - | 1 | 2309 | 8860423 |
Gene Symbol: CTAGE8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278507.1 | NP_001265436.1 | 1 | 2826 | ||
GenBank mRNA | AK292236.1 | - | 1 | 2330 |
Gene Symbol: CTAGE9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145659.1 | NP_001139131.1 | 1 | 2292 | ||
GenBank mRNA | DB518866.1 | - | 1 | 85 |
