siRNA Details
Gene Symbol: ZNF839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267827.1 | NP_001254756.1 | 2 | 791 | ||
NM_001267828.1 | NP_001254757.1 | 2 | 722 | |||
NM_018335.4 | NP_060805.3 | 2 | 804 | |||
XM_006720203.3 | XP_006720266.2 | 2 | 688 | |||
XM_011536945.1 | XP_011535247.1 | 3 | 906 | |||
XM_011536946.2 | XP_011535248.1 | 3 | 906 | |||
XM_011536948.2 | XP_011535250.1 | 2 | 532 | |||
XM_011536949.2 | XP_011535251.1 | 3 | 906 | |||
XM_011536950.2 | XP_011535252.1 | 3 | 906 | |||
XM_017021447.1 | XP_016876936.1 | 2 | 804 | |||
XM_017021448.1 | XP_016876937.1 | 2 | 804 | |||
XM_017021450.1 | XP_016876939.1 | 3 | 2987 | |||
XM_017021451.1 | XP_016876940.1 | 2 | 710 | |||
GenBank mRNA | AK093342.1 | - | 2 | 791 | ||
AK289632.1 | - | 2 | 722 | |||
AL137478.1 | - | 1 | 78 | |||
BX248266.1 | - | 2 | 804 | |||
BX367536.2 | - | 2 | 800 |
