siRNA Details
Gene Symbol: ZNF839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267827.1 | NP_001254756.1 | 5 | 1553 | ||
NM_001267828.1 | NP_001254757.1 | 5 | 1484 | |||
NM_018335.4 | NP_060805.3 | 5 | 1566 | |||
XM_006720203.3 | XP_006720266.2 | 5 | 1450 | |||
XM_011536945.1 | XP_011535247.1 | 6 | 1668 | |||
XM_011536948.2 | XP_011535250.1 | 5 | 1294 | |||
XM_011536949.2 | XP_011535251.1 | 6 | 1668 | |||
XM_017021448.1 | XP_016876937.1 | 5 | 1566 | |||
XM_017021449.1 | XP_016876938.1 | 4 | 832 | |||
XM_017021450.1 | XP_016876939.1 | 6 | 3749 | |||
XM_017021451.1 | XP_016876940.1 | 5 | 1472 | |||
GenBank mRNA | AF155112.1 | - | 3 | 195 | ||
AK093342.1 | - | 5 | 1553 | |||
AK127821.1 | - | 1 | 800 | |||
AK289632.1 | - | 5 | 1484 | |||
AL137478.1 | - | 4 | 840 | |||
BC073137.1 | - | 3 | 343 |
