siRNA Details
Gene Symbol: BSPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128326.2 | NP_001121798.1 | 4 | 296 | ||
XM_017026118.1 | XP_016881607.1 | 4 | 296 | |||
GenBank mRNA | DQ227497.1 | - | 4 | 297 |
