siRNA Details
Gene Symbol: TOMM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134493.1 | NP_001127965.1 | 1, 2 | 388 | ||
GenBank mRNA | AB435319.1 | - | 1, 2 | 133 | ||
AF216754.1 | - | 1, 2 | 159 | |||
AJ420506.1 | - | 1, 2 | 1097 | |||
BC015975.1 | - | 1, 2 | 170 | |||
BC018624.1 | - | 1, 2 | 152 | |||
BC022274.1 | - | 1, 2 | 146 | |||
BC107435.1 | - | 1, 2 | 164 | |||
BM783950.1 | - | 1, 2 | 498 | |||
L34839.1 | - | 1, 2 | 110 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
