siRNA Details
Gene Symbol: TOMM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134493.1 | NP_001127965.1 | 3 | 731 | ||
GenBank mRNA | AJ420506.1 | - | 3 | 1403 | ||
BC015975.1 | - | 3 | 514 | |||
BC018624.1 | - | 3 | 459 | |||
BC022274.1 | - | 3 | 453 | |||
BC107435.1 | - | 3 | 470 | |||
L34839.1 | - | 3 | 453 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
