siRNA Details
Gene Symbol: TOMM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134493.1 | NP_001127965.1 | 2 | 442 | ||
GenBank mRNA | AB435319.1 | - | 2 | 187 | ||
AF216754.1 | - | 2 | 213 | |||
AJ420506.1 | - | 2 | 1151 | |||
BC015975.1 | - | 2 | 224 | |||
BC018624.1 | - | 2 | 206 | |||
BC022274.1 | - | 2 | 200 | |||
BC107435.1 | - | 2 | 218 | |||
BM783950.1 | - | 2 | 552 | |||
L34839.1 | - | 2 | 164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
