siRNA Details
Gene Symbol: TNFRSF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004195.2 | NP_004186.1 | 1 | 46 | ||
NM_148901.1 | NP_683699.1 | 1 | 46 | |||
NM_148902.1 | NP_683700.1 | 1 | 46 | |||
XM_017002722.1 | XP_016858211.1 | 1 | 1237 | |||
GenBank mRNA | AY358877.1 | - | 1 | 28 | ||
BE244247.1 | - | 1 | 47 | |||
KP067911.1 | - | 1 | 46 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
