siRNA Details
Gene Symbol: CKLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040138.2 | NP_001035228.1 | 4 | 556 | ||
NM_016326.3 | NP_057410.1 | 2 | 498 | |||
NM_016951.3 | NP_058647.1 | 4 | 753 | |||
NM_181640.2 | NP_857591.1 | 3 | 594 | |||
NM_181641.2 | NP_857592.1 | 3 | 657 | |||
GenBank mRNA | AK290307.1 | - | 3 | 568 | ||
BU189908.1 | - | 3 | 554 | |||
DA113473.1 | - | 3 | 448 | |||
DA804889.1 | - | 4 | 537 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
