siRNA Details
Gene Symbol: FAM72B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100910.1 | NP_001094380.1 | 2 | 1047 | ||
NM_001320149.1 | NP_001307078.1 | 2 | 520 | |||
XM_011542018.1 | XP_011540320.1 | 3 | 1455 | |||
GenBank mRNA | BC046199.1 | - | 2 | 520 | 6064384 | |
BC137546.1 | - | 2 | 600 | 9021546 |
Gene Symbol: FAM72A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123168.2 | NP_001116640.1 | 2 | 1068 | ||
NM_001317901.1 | NP_001304830.1 | 2 | 288 | |||
NR_134239.1 | 3 | 348 | ||||
XM_006711526.3 | XP_006711589.1 | 3 | 1042 | |||
XM_011509965.2 | XP_011508267.1 | 3 | 951 | |||
XM_011509966.2 | XP_011508268.1 | 2 | 1083 | |||
XM_011509967.1 | XP_011508269.1 | 2 | 963 | |||
XM_017002233.1 | XP_016857722.1 | 3 | 948 | |||
GenBank mRNA | AW403341.1 | - | 3 | 331 | ||
BC035696.1 | - | 2 | 635 | 5541342 | ||
BC137545.1 | - | 2 | 599 | 9021545 | ||
BC146978.1 | - | 2 | 1067 | 9056906 | ||
BC146992.1 | - | 2 | 1066 | 9056920 | ||
BI916384.1 | - | 2 | 289 |
Gene Symbol: FAM72C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287385.1 | NP_001274314.1 | 2 | 1083 | ||
XM_011509723.2 | XP_011508025.1 | 3 | 1136 | |||
XM_011509724.2 | XP_011508026.1 | 2 | 1083 | |||
XM_017001682.1 | XP_016857171.1 | 3 | 1333 |
Gene Symbol: FAM72D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207418.2 | NP_997301.2 | 2 | 1066 | ||
XM_011509962.2 | XP_011508264.1 | 3 | 1101 | |||
XM_011509963.2 | XP_011508265.1 | 2 | 1083 | |||
XM_017002227.1 | XP_016857716.1 | 4 | 1486 | |||
GenBank mRNA | AB096683.1 | - | 2 | 1066 | ||
AK307197.1 | - | 2 | 1042 |

More Information
Other Species Targeted

Gene Ontology Categories:
