siRNA Details
Gene Symbol: FAM72B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100910.1 | NP_001094380.1 | 4 | 1543 | ||
NM_001320149.1 | NP_001307078.1 | 4 | 1016 | |||
XM_011542018.1 | XP_011540320.1 | 5 | 1951 | |||
GenBank mRNA | BC046199.1 | - | 4 | 1016 | 6064384 |
Gene Symbol: FAM72C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287385.1 | NP_001274314.1 | 4 | 1579 | ||
XM_011509723.2 | XP_011508025.1 | 5 | 1632 | |||
XM_017001682.1 | XP_016857171.1 | 5 | 1829 |
Gene Symbol: FAM72D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207418.2 | NP_997301.2 | 4 | 1562 | ||
XM_011509962.2 | XP_011508264.1 | 5 | 1597 | |||
XM_017002227.1 | XP_016857716.1 | 6 | 1982 | |||
GenBank mRNA | AB096683.1 | - | 4 | 1562 | ||
AK307197.1 | - | 3 | 1413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
