siRNA Details
Gene Symbol: FAM72B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100910.1 | NP_001094380.1 | 1 | 744 | ||
NM_001320149.1 | NP_001307078.1 | 1 | 337 | |||
GenBank mRNA | BC046199.1 | - | 1 | 337 | 6064384 | |
BC137546.1 | - | 1 | 297 | 9021546 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
