siRNA Details
Gene Symbol: HYI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190880.2 | NP_001177809.1 | 7 | 915 | ||
NM_001243526.1 | NP_001230455.1 | 8 | 990 | |||
NM_031207.5 | NP_112484.3 | 7 | 915 | |||
XM_005271239.4 | XP_005271296.1 | 8 | 1242 | |||
XM_005271240.4 | XP_005271297.1 | 7 | 1140 | |||
XM_006710937.3 | XP_006711000.1 | 8 | 1242 | |||
XM_011542222.2 | XP_011540524.1 | 7 | 1140 | |||
GenBank mRNA | AF284750.1 | - | 7 | 790 | ||
AF284751.1 | - | 7 | 790 | |||
AK096244.1 | - | 5 | 1991 | |||
AY037165.1 | - | 7 | 889 | |||
BC019041.2 | - | 7 | 717 | 4586546 | ||
JF911821.1 | - | 6 | 500 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
