siRNA Details
Gene Symbol: HYI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005271239.4 | XP_005271296.1 | 1 | 59 | ||
XM_005271240.4 | XP_005271297.1 | 1 | 59 | |||
XM_006710937.3 | XP_006711000.1 | 1 | 59 | |||
XM_011542222.2 | XP_011540524.1 | 1 | 59 | |||
GenBank mRNA | AF284750.1 | - | 1 | 71 | ||
AF284751.1 | - | 1 | 71 |
Gene Symbol: SZT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU633294.1 | - | 1 | 82 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
