siRNA Details
Gene Symbol: NSUN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168347.2 | NP_001161819.1 | 10 | 1425 | ||
NM_001168348.2 | NP_001161820.1 | 9 | 1386 | |||
NM_018044.4 | NP_060514.1 | 9 | 1500 | |||
NM_148956.3 | NP_683759.1 | 10 | 1430 | |||
XM_005277604.4 | XP_005277661.1 | 9 | 1473 | |||
GenBank mRNA | AF412028.1 | - | 10 | 1369 | ||
AF420249.1 | - | 9 | 1431 | |||
AK001129.1 | - | 9 | 1439 | |||
AK098619.1 | - | 10 | 1389 | |||
AK125667.1 | - | 9 | 2048 | |||
AK126375.1 | - | 9 | 1323 | |||
AK298221.1 | - | 10 | 1374 | |||
AK315361.1 | - | 10 | 1371 | |||
BC008084.2 | - | 10 | 1370 | 2823604 |
Gene Symbol: NSUN5P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033322.3 | 11 | 1536 | |||
NR_104013.1 | 8 | 1641 | ||||
GenBank mRNA | AF416610.1 | - | 10 | 1400 | ||
AK094312.1 | - | 4 | 2584 | |||
AK126486.1 | - | 6 | 4558 | |||
AK128374.1 | - | 4 | 2796 | |||
BC107112.1 | - | 3 | 660 | 40022630 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
