siRNA Details
Gene Symbol: IFNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021068.2 | NP_066546.1 | 1 | 597 | ||
GenBank mRNA | BC074965.2 | - | 1 | 549 | 30915507 | |
BC074966.2 | - | 1 | 549 | 30915288 | ||
BC113640.1 | - | 1 | 585 | 8322692 | ||
BC113642.1 | - | 1 | 585 | 8322694 | ||
M27318.1 | - | 1 | 669 |
Gene Symbol: IFNA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002171.2 | NP_002162.1 | 1 | 575 | ||
GenBank mRNA | BC069409.1 | - | 1 | 575 | 7262186 | |
BC103972.2 | - | 1 | 575 | 40015081 | ||
BC103973.1 | - | 1 | 575 | 40015082 | ||
V00551.1 | - | 1 | 575 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
