siRNA Details
Gene Symbol: IFNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021068.2 | NP_066546.1 | 1 | 238 | ||
GenBank mRNA | BC074965.2 | - | 1 | 190 | 30915507 | |
BC074966.2 | - | 1 | 190 | 30915288 | ||
BC113640.1 | - | 1 | 226 | 8322692 | ||
BC113642.1 | - | 1 | 226 | 8322694 | ||
M27318.1 | - | 1 | 310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
