siRNA Details
Gene Symbol: CCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123396.1 | NP_001116868.1 | 2 | 1585 | ||
XM_011534069.1 | XP_011532371.1 | 3 | 1298 | |||
GenBank mRNA | AK292685.1 | - | 2 | 1595 | ||
AK292920.1 | - | 2 | 1203 | |||
U03905.1 | - | 2 | 1180 | |||
X95583.1 | - | 1 | 440 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
