siRNA Details
Gene Symbol: FOXD4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012184.4 | NP_036316.1 | 1 | 1955 | ||
GenBank mRNA | AK122950.1 | - | 1 | 1955 | ||
AK125398.1 | - | 1 | 2374 | |||
AY344639.1 | - | 1 | 1867 |
Gene Symbol: FOXD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207305.4 | NP_997188.2 | 1 | 2070 | ||
GenBank mRNA | AY344640.1 | - | 1 | 1858 | ||
BC089432.1 | - | 1 | 2073 | 30555309 |
Gene Symbol: FOXD4L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199135.4 | NP_954586.4 | 1 | 1922 | ||
GenBank mRNA | AY344641.1 | - | 1 | 1922 |
Gene Symbol: FOXD4L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085476.1 | NP_001078945.1 | 1 | 2429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
