siRNA Details
Gene Symbol: FOXD4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012184.4 | NP_036316.1 | 1 | 1155 | ||
GenBank mRNA | AF452723.1 | - | 1 | 1003 | ||
AK122950.1 | - | 1 | 1155 | |||
AK125398.1 | - | 1 | 1574 | |||
AY344639.1 | - | 1 | 1067 | |||
BC136428.1 | - | 1 | 1147 | 9020417 |
Gene Symbol: FOXD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207305.4 | NP_997188.2 | 1 | 1270 | ||
GenBank mRNA | AF452724.1 | - | 1 | 250 | ||
AY344640.1 | - | 1 | 1058 | |||
BC089432.1 | - | 1 | 1273 | 30555309 | ||
BC103886.2 | - | 1 | 971 | |||
BC136570.1 | - | 1 | 1005 | 9020560 | ||
BC136571.1 | - | 1 | 1005 | 9020561 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
