siRNA Details
Gene Symbol: GAGE12J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098406.2 | NP_001091876.2 | 5 | 488 | ||
GenBank mRNA | BG186708.1 | - | 2 | 164 |
Gene Symbol: GAGE10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098413.3 | NP_001091883.3 | 5 | 478 |
Gene Symbol: GAGE12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127345.1 | NP_001120817.1 | 5 | 446 | ||
GenBank mRNA | BC130374.1 | - | 5 | 446 | 40146405 | |
BC143615.1 | - | 5 | 446 | 9052128 | ||
BC143619.1 | - | 5 | 446 | 9052132 |
Gene Symbol: GAGE12C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098408.1 | NP_001091878.1 | 5 | 481 |
Gene Symbol: GAGE12D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127199.1 | NP_001120671.1 | 5 | 492 |
Gene Symbol: GAGE12E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098418.1 | NP_001091888.1 | 5 | 481 |
Gene Symbol: GAGE12F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098405.2 | NP_001091875.1 | 5 | 515 |
Gene Symbol: GAGE12G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098409.1 | NP_001091879.1 | 5 | 481 |
Gene Symbol: GAGE12H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098410.1 | NP_001091880.1 | 5 | 481 |
Gene Symbol: GAGE13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098412.2 | NP_001091882.2 | 5 | 460 |
Gene Symbol: GAGE2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127212.1 | NP_001120684.1 | 5 | 459 | ||
GenBank mRNA | BC069309.1 | - | 5 | 434 | 7262139 | |
BC069397.1 | - | 5 | 434 | 7262151 | ||
BC069558.1 | - | 5 | 412 | 7262092 | ||
BG184057.1 | - | 2 | 159 |
Gene Symbol: GAGE2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U19143.1 | - | 5 | 445 |
Gene Symbol: GAGE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001476.1 | NP_001467.1 | 5 | 446 | ||
GenBank mRNA | U19147.1 | - | 5 | 446 |
Gene Symbol: GAGE7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC081536.1 | - | 4 | 310 | 30524801 | |
BC104951.1 | - | 5 | 446 | 8143954 |
Gene Symbol: GAGE8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012196.1 | NP_036328.1 | 5 | 434 | ||
GenBank mRNA | AF055473.1 | - | 5 | 434 | ||
BC143620.1 | - | 5 | 443 | 9052133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
