siRNA Details
Gene Symbol: KRTAP9-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031963.2 | NP_114169.2 | 1 | 942 |
Gene Symbol: KRTAP9-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031961.2 | NP_114167.2 | 1 | 960 | ||
GenBank mRNA | AJ406946.1 | - | 1 | 960 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
