siRNA Details
Gene Symbol: DAZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004081.5 | NP_004072.3 | 28 | 3435 | ||
XM_011531482.2 | XP_011529784.1 | 28 | 3473 | |||
XM_011531483.1 | XP_011529785.1 | 22 | 2600 | |||
GenBank mRNA | AK310133.1 | - | 4 | 1206 | ||
BC114927.1 | - | 21 | 1906 | 40028543 | ||
U66077.1 | - | 2 | 908 |
Gene Symbol: DAZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005785.2 | NP_001005785.1 | 23 | 2797 | ||
NM_001005786.2 | NP_001005786.2 | 16 | 2293 | |||
NM_020363.3 | NP_065096.2 | 24 | 2869 | |||
GenBank mRNA | AF414184.1 | - | 23 | 2779 | ||
AK302398.1 | - | 26 | 2936 | |||
AK302690.1 | - | 27 | 2938 | |||
BC113006.1 | - | 23 | 2605 | 40035862 |
Gene Symbol: DAZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020364.3 | NP_065097.2 | 19 | 2532 | ||
GenBank mRNA | BC113005.1 | - | 19 | 2317 | 40035860 |
Gene Symbol: DAZ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005375.2 | NP_001005375.1 | 23 | 2955 | ||
NM_020420.3 | NP_065153.1 | 17 | 2388 | |||
XM_005262602.1 | XP_005262659.1 | 29 | 3326 | |||
XM_011531509.2 | XP_011529811.1 | 29 | 3413 | |||
XM_011531510.1 | XP_011529812.1 | 23 | 2537 | |||
GenBank mRNA | AL833003.1 | - | 5 | 1266 | ||
BC047480.1 | - | 17 | 2293 | 5297459 | ||
BC047617.1 | - | 17 | 2289 | 4836748 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
