siRNA Details
Gene Symbol: ABHD17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130111.1 | NP_001123583.1 | 5 | 1538 | ||
NM_031213.3 | NP_112490.3 | 6 | 1691 | |||
XM_011528338.1 | XP_011526640.1 | 5 | 4563 | |||
XM_017027341.1 | XP_016882830.1 | 4 | 4410 | |||
XM_017027342.1 | XP_016882831.1 | 4 | 5667 | |||
GenBank mRNA | AK024419.1 | - | 3 | 4529 | ||
AK074043.1 | - | 4 | 4623 | |||
AK074548.1 | - | 5 | 1520 | |||
AK090438.1 | - | 3 | 5402 | |||
AK289533.1 | - | 5 | 1508 | |||
BC000158.2 | - | 3 | 1358 | 2985707 | ||
BC009256.2 | - | 5 | 1488 | 3954724 | ||
BC011667.2 | - | 5 | 1396 | 4122276 | ||
BC020512.1 | - | 6 | 1605 | 3850505 | ||
BC033749.1 | - | 5 | 1495 | 5197958 | ||
BC035961.1 | - | 5 | 1514 | 4805422 | ||
BC071644.1 | - | 5 | 1474 | 5242996 | ||
BC071876.1 | - | 5 | 1461 | 6672380 | ||
BC094816.1 | - | 5 | 1459 | 6266705 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
