siRNA Details
Gene Symbol: KRTAP10-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198699.1 | NP_941972.1 | 1 | 382 | ||
GenBank mRNA | AB076364.1 | - | 1 | 382 | ||
BC137339.1 | - | 1 | 382 | 9021337 | ||
BC137341.1 | - | 1 | 382 | 9021339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
