siRNA Details
Gene Symbol: KIR2DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015868.2 | NP_056952.2 | 8 | 1025 | ||
GenBank mRNA | AF022048.1 | - | 8 | 988 | ||
AF285435.1 | - | 7 | 948 | |||
AY789057.1 | - | 8 | 988 | |||
BC032422.1 | - | 8 | 1001 | 5220339 | ||
BC050730.1 | - | 8 | 1019 | 5219336 | ||
BC110613.1 | - | 8 | 1002 | 40031619 | ||
L41268.1 | - | 8 | 1025 | |||
L76662.1 | - | 8 | 999 | |||
L76663.1 | - | 8 | 999 | |||
U24074.1 | - | 8 | 1009 | |||
U50546.1 | - | 8 | 705 | |||
U73395.1 | - | 8 | 1009 |
Gene Symbol: KIR2DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014218.2 | NP_055033.2 | 8 | 1023 | ||
XM_017026782.1 | XP_016882271.1 | 8 | 1001 | |||
GenBank mRNA | AF022045.1 | - | 8 | 992 | ||
AF285431.1 | - | 8 | 1018 | |||
AF285432.1 | - | 7 | 945 | |||
AY366237.1 | - | 8 | 1021 | |||
AY366238.1 | - | 8 | 967 | |||
AY366239.1 | - | 8 | 1015 | |||
AY366240.1 | - | 8 | 1023 | |||
AY366241.1 | - | 8 | 1022 | |||
AY388471.1 | - | 8 | 1015 | |||
AY789055.1 | - | 9 | 1018 | |||
AY789056.1 | - | 8 | 985 | |||
BC069344.1 | - | 9 | 1096 | 7262296 | ||
KJ699235.1 | - | 8 | 1258 | |||
L41267.1 | - | 9 | 1017 | |||
U24076.1 | - | 9 | 1018 | |||
U24078.1 | - | 8 | 1025 |
Gene Symbol: KIR2DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014219.2 | NP_055034.2 | 8 | 1014 | ||
GenBank mRNA | AF285434.1 | - | 7 | 945 | ||
AY366242.1 | - | 8 | 1014 | |||
KP260924.1 | - | 8 | 981 | |||
L76669.1 | - | 8 | 996 | |||
U24075.1 | - | 8 | 999 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
