siRNA Details
Gene Symbol: CERS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024552.2 | NP_078828.2 | 9 | 1036 | ||
XM_011528290.2 | XP_011526592.1 | 9 | 1072 | |||
XM_011528291.2 | XP_011526593.1 | 9 | 1189 | |||
XM_011528292.2 | XP_011526594.1 | 10 | 2012 | |||
XM_011528293.2 | XP_011526595.1 | 10 | 1188 | |||
XM_011528294.2 | XP_011526596.1 | 10 | 1156 | |||
XM_011528295.2 | XP_011526597.1 | 8 | 842 | |||
XM_017027303.1 | XP_016882792.1 | 10 | 2044 | |||
XM_017027304.1 | XP_016882793.1 | 11 | 1267 | |||
XM_017027305.1 | XP_016882794.1 | 8 | 903 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
