siRNA Details
Gene Symbol: CGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033377.1 | NP_203695.2 | 3 | 630 | ||
XM_017026233.1 | XP_016881722.1 | 2 | 472 | |||
GenBank mRNA | BC107144.1 | - | 3 | 768 | ||
BC126460.1 | - | 3 | 512 |
Gene Symbol: CGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000737.3 | NP_000728.1 | 3 | 817 | ||
GenBank mRNA | AK291552.1 | - | 3 | 775 | ||
BC022796.1 | - | 3 | 755 | 4811294 | ||
BC069526.1 | - | 3 | 422 | 7262120 | ||
BC106059.1 | - | 3 | 763 | 4992804 | ||
BX334905.2 | - | 3 | 821 | |||
J00117.1 | - | 3 | 438 |
Gene Symbol: CGB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033043.1 | NP_149032.1 | 3 | 778 | ||
GenBank mRNA | BC006290.2 | - | 3 | 739 | 3960125 | |
BC051378.1 | - | 3 | 739 | 3956670 | ||
BC103970.1 | - | 3 | 509 | 40014677 | ||
BC103971.1 | - | 3 | 505 | 40014679 | ||
BC106723.1 | - | 3 | 423 | 40014469 | ||
BC106724.1 | - | 3 | 422 | 40014470 | ||
BT006890.1 | - | 2 | 413 |
Gene Symbol: CGB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033183.2 | NP_149439.1 | 3 | 785 | ||
GenBank mRNA | BC030994.1 | - | 3 | 780 | 4605021 | |
BC103969.1 | - | 3 | 505 | 40014676 | ||
BC128603.1 | - | 3 | 505 | 40002893 | ||
BX380841.2 | - | 3 | 767 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
