siRNA Details
Gene Symbol: TSTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113205.1 | NP_001106676.1 | 3 | 514 | ||
NM_001113206.1 | NP_001106677.1 | 3 | 305 | |||
NM_001113207.1 | NP_001106678.1 | 4 | 428 | |||
GenBank mRNA | BC001699.1 | - | 3 | 460 | ||
BC038219.1 | - | 3 | 460 | |||
CD675389.1 | - | 4 | 423 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
