siRNA Details
Gene Symbol: CERS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290265.1 | NP_001277194.1 | 6 | 1913 | ||
NM_198207.2 | NP_937850.1 | 6 | 2067 | |||
GenBank mRNA | BC084582.1 | - | 6 | 1865 | 6192817 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
