siRNA Details
Gene Symbol: HIST1H4I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003495.2 | NP_003486.1 | 1 | 315 | ||
GenBank mRNA | BC016336.1 | - | 1 | 346 | ||
BC066250.1 | - | 1 | 315 | 7002105 | ||
BC067496.1 | - | 1 | 315 | 7002103 | ||
BC067497.1 | - | 1 | 315 | 7002106 | ||
BC075806.1 | - | 1 | 327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
