siRNA Details
Gene Symbol: POLR1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206559.1 | NP_001193488.1 | 3 | 1444 | ||
NM_152705.2 | NP_689918.1 | 3 | 1277 | |||
XM_005266414.2 | XP_005266471.1 | 4 | 2117 | |||
XM_017020622.1 | XP_016876111.1 | 2 | 1144 | |||
XM_017020623.1 | XP_016876112.1 | 4 | 2037 | |||
GenBank mRNA | AK097973.1 | - | 4 | 1363 | ||
AK307356.1 | - | 3 | 1241 | |||
BC006972.1 | - | 3 | 1177 | |||
BC015319.2 | - | 3 | 1186 | 3847651 | ||
BC018528.1 | - | 3 | 1190 | 3865616 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
