siRNA Details
Gene Symbol: POLR1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206559.1 | NP_001193488.1 | 3 | 558 | ||
NM_152705.2 | NP_689918.1 | 3 | 391 | |||
XM_005266414.2 | XP_005266471.1 | 4 | 1231 | |||
XM_017020622.1 | XP_016876111.1 | 2 | 258 | |||
XM_017020623.1 | XP_016876112.1 | 4 | 1151 | |||
GenBank mRNA | AF131855.1 | - | 1 | 1144 | ||
AK097973.1 | - | 4 | 478 | |||
AK307356.1 | - | 3 | 355 | |||
AK311833.1 | - | 3 | 278 | |||
BC006972.1 | - | 3 | 291 | |||
BC015319.2 | - | 3 | 300 | 3847651 | ||
BC018528.1 | - | 3 | 304 | 3865616 | ||
DB455307.1 | - | 3 | 391 | |||
DB459107.1 | - | 3 | 348 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
