siRNA Details
Gene Symbol: RPS18P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077247.1 | 1 | 745 | |||
GenBank mRNA | BC071678.1 | - | 1 | 389 | ||
EB387363.1 | - | 1 | 745 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
