siRNA Details
Gene Symbol: C1orf204 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134233.1 | NP_001127705.1 | 6 | 1707 | ||
GenBank mRNA | AK096506.1 | - | 6 | 1707 | ||
AL080176.1 | - | 1 | 114 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
